1-Ethyl-4-nitrobenzene ≥99.0%

Supplier: TCI America
Warning

Synonyms: 4-Ethylnitrobenzene

E0154-500G E0154-25G
TCE0154-500GEA 246.48 CAD
TCE0154-500G TCE0154-025G
1-Ethyl-4-nitrobenzene ≥99.0%
1-Ethyl-4-nitrobenzene

CAS Number: 100-12-9
MDL Number: MFCD00007385
Molecular Formula: C8H9NO2
Molecular Weight: 151.17
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 242
Melting point (°C): -18
Flash Point (°C): 117
Specific Gravity (20/20): 1.12

Formula: C₈H₉NO₂
MW: 151.16 g/mol
Boiling Pt: 242 °C
Melting Pt: –18 °C
Density: 1.12 (20 °C)
Flash Pt: 117 °C
MDL Number: MFCD00007385
CAS Number: 100-12-9
EINECS: 202-821-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3946136f52eb8df4480d9cca847d98048cc458c1&vid=342728d1e61f16718a7099da2df5b342a5a71661&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR