1-Ethyl-2-nitrobenzene ≥99.0% for synthesis

Supplier: TCI America

Synonyms: 2-Ethylnitrobenzene

E0153-25G E0153-500G
TCE0153-025GEA 51.44 CAD
TCE0153-025G TCE0153-500G
1-Ethyl-2-nitrobenzene ≥99.0% for synthesis
1-Ethyl-2-nitrobenzene

CAS Number: 612-22-6
MDL Number: MFCD00024294
Molecular Formula: C8H9NO2
Molecular Weight: 151.17
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 232
Melting point (°C): -13
Flash Point (°C): 109
Specific Gravity (20/20): 1.12

Formula: C₈H₉NO₂
Boiling Pt: 232 °C
Melting Pt: –13 °C
Density: 1.12 (20 °C)
Flash Pt: 109 °C
MDL Number: MFCD00024294
CAS Number: 612-22-6
EINECS: 210-299-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a16fc096d537bd8cd35e95334092cdf232f1316b&vid=4228a3e19cefffc503d1f54e8b388c31567d14bd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR