N-Ethyl-1-naphthylamine hydrobromide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

E0149-25G E0149-5G
TCE0149-025GEA 229.1 CAD
TCE0149-025G TCE0149-5G
N-Ethyl-1-naphthylamine hydrobromide ≥98.0% (by titrimetric analysis)
N-Ethyl-1-naphthylamine hydrobromide

CAS Number: 36966-04-8
MDL Number: MFCD00060154
Molecular Formula: C12H13N
Molecular Weight: 252.16
Purity/Analysis Method: >98.0% (T)
Form: Crystal

Formula: C₁₂H₁₃N
MDL Number: MFCD00060154
CAS Number: 36966-04-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db11ec05ad05d08903a08d915b062afc9dcc7774&vid=db11ec05ad05d08903a08d915b062afc9dcc7774&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR