7-Ethyl-2-methyl-4-undecanol ≥98.0%

Supplier: TCI America

Synonyms: Tetradecyl Alcohol, Tetradecanol, 7-Ethyl-2-methylundecan-4-ol

E0144-25ML E0144-500ML
TCE0144-025MLEA 109.02 CAD
TCE0144-025ML TCE0144-500ML
7-Ethyl-2-methyl-4-undecanol ≥98.0%
7-Ethyl-2-methyl-4-undecanol

CAS Number: 103-20-8
MDL Number: MFCD00027253
Molecular Formula: C14H30O
Molecular Weight: 214.39
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 264
Flash Point (°C): 141
Specific Gravity (20/20): 0.84


Other Product Names / Synonyms: Tetradecanol, Tetradecyl Alcohol

Formula: C₁₄H₃₀O
MW: 214.39 g/mol
Boiling Pt: 264 °C
Density: 0.84 (20 °C)
Flash Pt: 141 °C
MDL Number: MFCD00027253
CAS Number: 103-20-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1478450599c78b66d9711e630b2938536cd1a4aa&vid=1478450599c78b66d9711e630b2938536cd1a4aa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR