Tris(trifluoro-2,4-pentanedionato)aluminum(III) ≥98.0%

Supplier: TCI America

Synonyms: Aluminum(III) Trifluoroacetylacetonate, Trifluoroacetylacetonato Aluminum(III) Salt

A1267-5G A1267-25G
TCA1267-005GEA 175.22 CAD
TCA1267-005G TCA1267-25G
Tris(trifluoro-2,4-pentanedionato)aluminum(III) ≥98.0%
Tris(trifluoro-2,4-pentanedionato)aluminum(III)

CAS Number: 14354-59-7
MDL Number: MFCD00042062
Molecular Formula: C15H12AlF9O6
Molecular Weight: 486.22
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 121


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts
Other Product Names / Synonyms: Aluminum(III) Trifluoroacetylacetonate, Trifluoroacetylacetonato Aluminum(III) Salt

Formula: C₁₅H₁₂AlF₉O₆
MW: 486.22 g/mol
Melting Pt: 121 °C
MDL Number: MFCD00042062
CAS Number: 14354-59-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f56cc96ac33d7bababde9ba9b201d2a6861b56a&vid=3f56cc96ac33d7bababde9ba9b201d2a6861b56a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR