1,2-Bis(chloroacetoxy)ethane ≥98.0%

Supplier: TCI America

Synonyms: Ethyleneglycolbis(monochloroacetatE), Ethylene Glycol Dichloroacetate, Ethylene Glycol Bis(monochloroacetate)

E0107-25G E0107-500G
TCE0107-025GEA 52.14 CAD
TCE0107-025G TCE0107-500G
1,2-Bis(chloroacetoxy)ethane ≥98.0%
1,2-Bis(chloroacetoxy)ethane

CAS Number: 6941-69-1
MDL Number: MFCD00045277
Molecular Formula: C6H8Cl2O4
Molecular Weight: 215.03
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 158
Melting point (°C): 41


Other Product Names / Synonyms: Ethylene Glycol Bis(monochloroacetate), Ethylene Glycol Dichloroacetate

Formula: C₆H₈Cl₂O₄
MW: 215.03 g/mol
Boiling Pt: 158 °C
Melting Pt: 41 °C
MDL Number: MFCD00045277
CAS Number: 6941-69-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82f34b4c8dc71d97cfc538876059fa0b1de14bdb&vid=82f34b4c8dc71d97cfc538876059fa0b1de14bdb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR