EDTA tetrasodium salt dihydrate ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

E0099-25G E0099-500G
TCE0099-25GEA 35.99 CAD
TCE0099-25G TCE0099-500G
EDTA tetrasodium salt dihydrate ≥98.0% (by titrimetric analysis)
EDTA tetrasodium salt dihydrate

CAS Number: 10378-23-1
MDL Number: MFCD00012460
Molecular Formula: C10H16N2O8
Molecular Weight: 380.17
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 300


Product Categories: Analytical Science, Complexones (Chelating Agents)
Other Product Names / Synonyms: Ethylenediaminetetraacetic Acid Tetrasodium Salt, EDTA 4Na

Formula: C₁₀H₁₂N₂Na₄O₈·2H₂O
Melting Pt: 300 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00150026
CAS Number: 10378-23-1
EINECS: 200-573-9
Merck Index: 13,03544
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=648457ec2ece56b46f802879799d86251e150e38&vid=648457ec2ece56b46f802879799d86251e150e38&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR