Copper(II) disodium ethylenediaminetetraacetate tetrahydrate ≥98.0%

Supplier: TCI America

Synonyms: Ethylenediaminetetraacetic acid Copper(II) disodium salt, Copper(II) disodium 2,2',2'',2'''-(ethane-1,2-diylbis(azanetriyl))tetraacetate tetrahydrate, Copper Disodium Ethylenediaminetetraacetate hydrate

E0090-25G E0090-500G
TCE0090-025GEA 50.56 CAD
TCE0090-025G TCE0090-500G
Copper(II) disodium ethylenediaminetetraacetate tetrahydrate ≥98.0%
Copper(II) disodium ethylenediaminetetraacetate tetrahydrate

CAS Number: 39208-15-6
MDL Number: MFCD00148920
Molecular Formula: C10H16N2O8
Molecular Weight: 397.74
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Other Product Names / Synonyms: Ethylenediaminetetraacetic Acid Copper(II) Disodium Salt

Formula: C₁₀H₁₂CuN₂Na₂O₈·4H₂O
MW: 469.80 g/mol
MDL Number: MFCD00148920
CAS Number: 39208-15-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=429b12a5fd81b7edd12e4c5d6d2e567d1812ba97&vid=429b12a5fd81b7edd12e4c5d6d2e567d1812ba97&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR