4-Ethoxychrysoidine hydrochloride ≥95.0% (by titrimetric analysis)

Supplier: TCI America

E0054-5G E0054-25G
TCE0054-5GEA 372.49 CAD
TCE0054-5G TCE0054-025G
4-Ethoxychrysoidine hydrochloride ≥95.0% (by titrimetric analysis)
4-Ethoxychrysoidine hydrochloride

CAS Number: 2313-87-3
MDL Number: MFCD00042023
Molecular Formula: C14H16N4O
Molecular Weight: 292.77
Purity/Analysis Method: >95.0% (T)
Form: Crystal


Product Categories: Analytical Science, Indicators (pH)
Other Product Names / Synonyms: 4-(4-Ethoxyphenylazo)-1,3-phenylenediamine Monohydrochloride, Ethoxy Red

Formula: C₁₄H₁₆N₄O
MDL Number: MFCD00042023
CAS Number: 2313-87-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1af68d055b8d629baf8c43109789e7ef36e3d92f&vid=1af68d055b8d629baf8c43109789e7ef36e3d92f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR