Ethyl (ethoxymethylene)acetate ≥98.0% for synthesis
Supplier: TCI America
Synonyms:
3-Ethoxyacrylic acid ethyl ester, Ethyl 3-Ethoxyacrylate, Ethyl 3-Ethoxy-2-Propenoate, Ethyl 3-ethoxyacrylate
E0041-5ML
E0041-25ML
TCE0041-005MLEA
86.9
CAD
TCE0041-005ML
TCE0041-025ML
Ethyl (ethoxymethylene)acetate ≥98.0% for synthesis
Ethyl (ethoxymethylene)acetate
CAS Number: 1001-26-9
MDL Number: MFCD00009863
Molecular Formula: C7H12O3
Molecular Weight: 144.17
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 76
Specific Gravity (20/20): 1.00
Other Product Names / Synonyms: 3-Ethoxyacrylic Acid Ethyl Ester
Formula:
C₇H₁₂O₃ MW: 144.17 g/mol Density: 1.00 (20 °C) Flash Pt: 76 °C |
MDL Number:
MFCD00009863 CAS Number: 1001-26-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...