Esculin hydrate ≥98.0% (by HPLC, titration analysis), crystals

Supplier: TCI America

Synonyms: Aesculin, 6,7-Dihydroxycoumarin 6-glucoside hydrate, Esculin

E0024-5G E0024-25G
TCE0024-005GEA 126.1 CAD
TCE0024-005G TCE0024-025G
Esculin hydrate ≥98.0% (by HPLC, titration analysis), crystals
Esculin hydrate

CAS Number: 531-75-9 MDL Number: MFCD00006879 Molecular Formula: C15H16O9 Molecular Weight: 340.28 Purity/Analysis Method: 98.0% (HPLC,T) Form: Crystal Melting point (°C): 206 Specific rotation [a]20/D: -79 deg (C=2.4, Dioxane(50%))


Specific rotation [a]20/D: -79 deg (C=2.4, Dioxane(50%))
Product Categories: Bioscience, Glycosides, Glucose, Glycosides, Phenylpropanoids, Phenylpropanoids (Phytochemicals)
Other Product Names / Synonyms: 7-Hydroxycoumarin-6-yl beta-D-Glucopyranoside

Formula: C₁₅H₁₆O₉
MW: 340.29 g/mol (anhy)
Melting Pt: 206 °C
MDL Number: MFCD00149492
CAS Number: 531-75-9
EINECS: 208-517-5
Merck Index: 12,03739
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ff2048f42d38fa999b657ff9b18a172a49e6f9b8&vid=69dc27ea469a8eda3218a7c086b1e8c90c19479f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR