Disodium pamidronate, hydrate ≥97.0% (by titrimetric analysis)
Supplier: TCI America
D3921-100MG
D3921-1G
TCD3921-100MGEA
83.74
CAD
TCD3921-100MG
TCD3921-1G
Disodium pamidronate, hydrate ≥97.0% (by titrimetric analysis)
Disodium pamidronate
CAS Number: 57248-88-1
MDL Number: MFCD11045314
Molecular Formula: C3H11NO7P2
Molecular Weight: 279.03
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Storage Temperature: 0-10°C
Product Categories: Bioscience, Anti-osteoporosis Ingredients
Other Product Names / Synonyms: Pamidronic Acid Disodium Salt, Disodium 3-Amino-1-hydroxypropane-1,1-diphosphonate
Formula:
C₃H₉NO₇P₂Na₂ |
MDL Number:
MFCD11045314 CAS Number: 57248-88-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...