Diisopropyl fumarate ≥98.0%
Supplier: TCI America
Synonyms:
Fumaric Acid Diisopropyl Ester
D3916-25G
D3916-500G
TCD3916-25GEA
55.3
CAD
TCD3916-25G
TCD3916-500G
Diisopropyl fumarate ≥98.0%
Diisopropyl fumarate
CAS Number: 7283-70-7
MDL Number: MFCD00545172
Molecular Formula: C10H16O4
Molecular Weight: 200.23
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 110
Melting point (°C): 2
Specific Gravity (20/20): 0.99
Product Categories: Materials Chemistry, Copolymerization Monomers
Other Product Names / Synonyms: Fumaric Acid Diisopropyl Ester
Formula:
C₁₀H₁₆O₄ Boiling Pt: 110 °C Melting Pt: 2 °C Density: 0.99 (20 °C) |
MDL Number:
MFCD00545172 CAS Number: 7283-70-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...