Diisobutyl fumarate ≥98.0%
Supplier: TCI America
D3915-25G
D3915-500G
TCD3915-25GEA
63.2
CAD
TCD3915-25G
TCD3915-500G
Diisobutyl fumarate ≥98.0%
Diisobutyl fumarate
CAS Number: 7283-69-4
MDL Number: MFCD00152430
Molecular Formula: C12H20O4
Molecular Weight: 228.29
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 122
Melting point (°C): 8
Specific Gravity (20/20): 0.98
Product Categories: Materials Chemistry, Copolymerization Monomers
Other Product Names / Synonyms: Fumaric Acid Diisobutyl Ester
Formula:
C₁₂H₂₀O₄ Boiling Pt: 122 °C Melting Pt: 8 °C Density: 0.98 (20 °C) |
MDL Number:
MFCD00152430 CAS Number: 7283-69-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...