2-Hydroxy-4-methoxyaniline hydrochloride ≥98.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: 2-Hydroxy-p-anisidine Hydrochloride, 2-Amino-5-methoxyphenol Hydrochloride

A1259-5G A1259-1G
TCA1259-5GEA 466.1 CAD
TCA1259-5G TCA1259-001G
2-Hydroxy-4-methoxyaniline hydrochloride ≥98.0% (by total nitrogen basis)
2-Hydroxy-4-methoxyaniline hydrochloride

CAS Number: 39547-15-4
MDL Number: MFCD00052002
Molecular Formula: C7H9NO2
Molecular Weight: 175.61
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 213


Other Product Names / Synonyms: 2-Amino-5-methoxyphenol Hydrochloride, 2-Hydroxy-p-anisidine Hydrochloride

Formula: C₇H₉NO₂
MW: 175.61 g/mol
Melting Pt: 213 °C
MDL Number: MFCD00052002
CAS Number: 39547-15-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=91eaee106a0bf2ccee0f772c85482b3b54ff95bc&vid=91eaee106a0bf2ccee0f772c85482b3b54ff95bc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR