4,4'-Dimethoxy-2,2'-bipyridyl ≥98.0%

Supplier: TCI America

Synonyms: 4,4'-Dimethoxy-2,2'-bipyridine

D3886-1G D3886-5G
TCD3886-1GEA 105.86 CAD
TCD3886-1G TCD3886-5G
4,4'-Dimethoxy-2,2'-bipyridyl ≥98.0%
4,4'-Dimethoxy-2,2'-bipyridyl

CAS Number: 17217-57-1
MDL Number: MFCD00233880
Molecular Formula: C12H12N2O2
Molecular Weight: 216.24
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 172

Formula: C₁₂H₁₂N₂O₂
MW: 216.24 g/mol
Melting Pt: 172 °C
MDL Number: MFCD00233880
CAS Number: 17217-57-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3dce0c144bdde0dd31cdb1e496d8a65d09f51d1d&vid=3dce0c144bdde0dd31cdb1e496d8a65d09f51d1d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR