Diphenyl-4-bromo-ɑ-chlorobenzylphosphonate ≥95.0% (by titrimetric analysis)
Supplier: TCI America
D3875-5G
D3875-25G
TCD3875-5GEA
325.64
CAD
TCD3875-5G
TCD3875-25G
Diphenyl-4-bromo-ɑ-chlorobenzylphosphonate ≥95.0% (by titrimetric analysis)
Diphenyl-4-bromo-alpha-chlorobenzylphosphonate
CAS Number: 189099-56-7
Molecular Formula: C19H15BrClO3P
Molecular Weight: 437.65
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 107
Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: 4-Bromo-alpha-chlorobenzylphosphonic Acid Diphenyl Ester
Formula:
C₁₉H₁₅BrClO₃P Melting Pt: 107 °C |
CAS Number:
189099-56-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...