Diethyl (methoxymethyl)phosphonate ≥98.0%

Supplier: TCI America

D3873-25G D3873-5G
TCD3873-25GEA 541.94 CAD
TCD3873-25G TCD3873-5G
Diethyl (methoxymethyl)phosphonate ≥98.0%
Diethyl (methoxymethyl)phosphonate

CAS Number: 32806-04-5
Molecular Formula: C6H15O4P
Molecular Weight: 182.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 88
Specific Gravity (20/20): 1.09


Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: (Methoxymethyl)phosphonic Acid Diethyl Ester

Formula: C₆H₁₅O₄P
Boiling Pt: 88 °C
Density: 1.09 (20 °C)
CAS Number: 32806-04-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d28961aff04e75e1ac796b4a38935321d09c0699&vid=d28961aff04e75e1ac796b4a38935321d09c0699&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR