1,3-Dimesitylimidazol-2-ylidene ≥97.0%
Supplier: TCI America
Synonyms:
1,3-Bis(2,4,6-trimethylphenyl)imidazol-2-ylidene, IMes, 1,3-Bis(2,4,6-trimethylphenyl)-1,3-dihydro-2H-imidazol-2-ylidene
CAS Number: 141556-42-5
MDL Number: MFCD09265338
Molecular Formula: C21H24N2
Molecular Weight: 304.44
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 155
Storage Temperature: <0°C
Product Categories: Synthetic Organic Chemistry, Cross-coupling Reaction using Transition Metal Catalysts, Ligands, N-Heterocyclic Carbene Ligands ([2+2+2] Cycloaddition Reaction)
Other Product Names / Synonyms: IMes
Formula:
C₂₁H₂₄N₂ Melting Pt: 155 °C |
MDL Number:
MFCD09265338 CAS Number: 141556-42-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...