Carbazochrome ≥99.0% (by HPLC)

Supplier: TCI America

A0176-5G A0176-25G
TCA0176-005GEA 36.34 CAD
TCA0176-005G TCA0176-025G
Carbazochrome ≥99.0% (by HPLC)
Carbazochrome

CAS Number: 69-81-8
MDL Number: MFCD00059705
Molecular Formula: C10H12N4O3
Molecular Weight: 236.23
Purity/Analysis Method: >99.0% (HPLC)
Form: Crystal
Melting point (°C): 203


Other Product Names / Synonyms: Adrenochrome Semicarbazone

Formula: C₁₀H₁₂N₄O₃
Melting Pt: 203 °C
MDL Number: MFCD00059705
CAS Number: 69-81-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=099e6980c278aaf4882af91616f708808ff357fd&vid=099e6980c278aaf4882af91616f708808ff357fd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR