2,7-Dibromo-9,9-dimethylfluorene ≥98.0%

Supplier: TCI America

Synonyms: 2,7-Dibromo-9,9-dimethyl-9H-fluorene

D3859-25G D3859-5G
TCD3859-25GEA 609.88 CAD
TCD3859-25G TCD3859-5G
2,7-Dibromo-9,9-dimethylfluorene ≥98.0%
2,7-Dibromo-9,9-dimethylfluorene

CAS Number: 28320-32-3
MDL Number: MFCD00099471
Molecular Formula: C15H12Br2
Molecular Weight: 352.07
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 179

Formula: C₁₅H₁₂Br₂
MW: 352.07 g/mol
Melting Pt: 179 °C
MDL Number: MFCD00099471
CAS Number: 28320-32-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bbf567f21f0eed0b78e6ff4996b7a3294d8d68c9&vid=bbf567f21f0eed0b78e6ff4996b7a3294d8d68c9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR