trans-4-(3,4-Difluorophenyl)-trans-4'-ethylbicyclohexane ≥97.0%
Supplier: TCI America
Synonyms:
trans,trans-4-(3,4-Difluorophenyl)-4'-ethylbicyclohexyl
D3856-5G
D3856-1G
TCD3856-5GEA
150.1
CAD
TCD3856-5G
TCD3856-1G
trans-4-(3,4-Difluorophenyl)-trans-4'-ethylbicyclohexane ≥97.0%
trans-4-(3,4-Difluorophenyl)-trans-4'-ethylbicyclohexane
CAS Number: 118164-50-4
MDL Number: MFCD11053389
Molecular Formula: C20H28F2
Molecular Weight: 306.44
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 53
Other Product Names / Synonyms: trans,trans-4-(3,4-Difluorophenyl)-4'-ethylbicyclohexyl
Formula:
C₂₀H₂₈F₂ MW: 306.44 g/mol Melting Pt: 53 °C |
MDL Number:
MFCD11053389 CAS Number: 118164-50-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...