4,4'-Di(1,2,3-triazolyl) Disulfide Hydrate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
D3855-1G
D3855-5G
TCD3855-1GEA
229.73
CAD
TCD3855-1G
TCD3855-5G
4,4'-Di(1,2,3-triazolyl) Disulfide Hydrate ≥98.0% (by HPLC, titration analysis)
4,4'-Di(1,2,3-triazolyl) Disulfide Hydrate
CAS Number: 6440-09-1
Molecular Formula: C4H4N6S2
Molecular Weight: 200.24
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Other Product Names / Synonyms: 4,4'-Dithiobis(1,2,3-triazole)
Formula:
C₄H₄N₆S₂ |
CAS Number:
6440-09-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...