2,2'-Dipyrrolylmethane ≥97.0%
Supplier: TCI America
D3851-1G
D3851-5G
TCD3851-1GEA
299.36
CAD
TCD3851-1G
TCD3851-5G
2,2'-Dipyrrolylmethane ≥97.0%
2,2'-Dipyrrolylmethane
CAS Number: 21211-65-4
MDL Number: MFCD03931560
Molecular Formula: C9H10N2
Molecular Weight: 146.19
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 120
Melting point (°C): 74
Lambda max.: 449 nm (CHCl3)Storage Temperature: 0-10°C
Formula:
C₉H₁₀N₂ Boiling Pt: 120 °C Melting Pt: 74 °C |
MDL Number:
MFCD03931560 CAS Number: 21211-65-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...