1,3-Di-tert-butyl-1,3,2-diazaphospholidine-2-oxide ≥95.0%

Supplier: TCI America

D3846-1G D3846-5G
TCD3846-1GEA 180.12 CAD
TCD3846-1G TCD3846-5G
1,3-Di-tert-butyl-1,3,2-diazaphospholidine-2-oxide ≥95.0%
1,3-Di-tert-butyl-1,3,2-diazaphospholidine-2-oxide

CAS Number: 854929-38-7
Molecular Formula: C10H23N2OP
Molecular Weight: 218.28
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 82

Formula: C₁₀H₂₃N₂OP
MW: 218.28 g/mol
Melting Pt: 82 °C
CAS Number: 854929-38-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eab6bd6f6d6296dcf2ff1871b6f660e19455b985&vid=eab6bd6f6d6296dcf2ff1871b6f660e19455b985&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR