1-[2-(Di-tert-butylphosphino)phenyl]-3,5-diphenyl-1H-pyrazole ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D3834-5G D3834-1G
TCD3834-5GEA 377.62 CAD
TCD3834-5G TCD3834-1G
1-[2-(Di-tert-butylphosphino)phenyl]-3,5-diphenyl-1H-pyrazole ≥98.0% (by GC, titration analysis)
1-[2-(Di-tert-butylphosphino)phenyl]-3,5-diphenyl-1H-pyrazole

CAS Number: 628333-86-8
MDL Number: MFCD09038823
Molecular Formula: C29H33N2P
Molecular Weight: 440.57
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 142

Formula: C₂₉H₃₃N₂P
Melting Pt: 142 °C
MDL Number: MFCD09038823
CAS Number: 628333-86-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f68a3a7272428e8bf573c82e60e877507eefc3dd&vid=f68a3a7272428e8bf573c82e60e877507eefc3dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR