4-(N,N-Dimethylamino)phenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 4-(Dimethylamino)phenylboronic acid pinacol ester, N,N-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

D3832-1G D3832-5G
TCD3832-1GEA 72.68 CAD
TCD3832-1G TCD3832-5G
4-(N,N-Dimethylamino)phenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
4-(N,N-Dimethylamino)phenylboronic acid pinacol ester

CAS Number: 171364-78-6
MDL Number: MFCD05663854
Molecular Formula: C14H22BNO2
Molecular Weight: 247.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 125


Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives
Other Product Names / Synonyms: 2-[4-(Dimethylamino)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-(Dimethylamino)phenylboronic Acid Pinacol Ester

Formula: C₁₄H₂₂BNO₂
MW: 247.1410 g/mol
Melting Pt: 125 °C
MDL Number: MFCD05663854
CAS Number: 171364-78-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=680b3be34733352ba203f196772f1eedec5cf083&vid=680b3be34733352ba203f196772f1eedec5cf083&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR