Diethylene glycol-2-bromoethyl methyl ether ≥94.0%

Supplier: TCI America

Synonyms: 1-Bromo-3,6,9-trioxadecane, 2-[2-(2-Methoxyethoxy)ethoxy]ethyl bromide, Diethylene Glycol 2-Bromoethyl Methyl Ether, mPEG3-bromide, Methyl-PEG3-bromide

D3831-5G D3831-25G
TCD3831-5GEA 205.59 CAD
TCD3831-5G TCD3831-25G
Diethylene glycol-2-bromoethyl methyl ether ≥94.0%
Diethylene glycol-2-bromoethyl methyl ether

CAS Number: 72593-77-2
Molecular Formula: C7H15BrO3
Molecular Weight: 227.10
Purity/Analysis Method: >94.0% (GC)
Form: Clear Liquid
Boiling point (°C): 65
Specific Gravity (20/20): 1.31
Storage Temperature: <0°C


Product Categories: Bioscience, PEGylation Reagents & PEG Spacers, PEGylation Reagents
Other Product Names / Synonyms: 2-[2-(2-Methoxyethoxy)ethoxy]ethyl Bromide, 1-Bromo-3,6,9-trioxadecane, mPEG3-Bromide

Formula: C₇H₁₅BrO₃
MW: 227.10 g/mol
Boiling Pt: 65 °C
Density: 1.31 (20 °C)
MDL Number: MFCD00956116
CAS Number: 72593-77-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c97afcf3a23618276b607e4bf76b45ad61fb9bca&vid=c97afcf3a23618276b607e4bf76b45ad61fb9bca&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 94.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR