Diphenyl-ɑ-chlorobenzylphosphonate ≥98.0%

Supplier: TCI America

Synonyms: ɑ-Chlorobenzylphosphonic Acid Diphenyl Ester

D3824-5G D3824-1G
TCD3824-5GEA 543.84 CAD
TCD3824-5G TCD3824-1G
Diphenyl-ɑ-chlorobenzylphosphonate ≥98.0%
Diphenyl-alpha-chlorobenzylphosphonate

CAS Number: 58263-67-5
Molecular Formula: C19H16ClO3P
Molecular Weight: 358.76
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Boiling point (°C): 210


Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: alpha-Chlorobenzylphosphonic Acid Diphenyl Ester

Formula: C₁₉H₁₆ClO₃P
Boiling Pt: 210 °C
CAS Number: 58263-67-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d545ccc95350a9ba85fcf5379d4de5209a70266e&vid=d545ccc95350a9ba85fcf5379d4de5209a70266e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR