(S)-1-(Diphenylphosphino)-2-[(S)-4-isopropyloxazolin-2-yl]ferrocene ≥97.0% (by GC, titration analysis)

Supplier: TCI America

D3822-1G D3822-200MG
TCD3822-1GEA 714.16 CAD
TCD3822-1G TCD3822-200MG
(S)-1-(Diphenylphosphino)-2-[(S)-4-isopropyloxazolin-2-yl]ferrocene ≥97.0% (by GC, titration analysis)
(S)-1-(Diphenylphosphino)-2-[(S)-4-isopropyloxazolin-2-yl]ferrocene

CAS Number: 163169-29-7
MDL Number: MFCD15144869
Molecular Formula: C28H28FeNOP
Molecular Weight: 481.36
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 160


Product Categories: Synthetic Organic Chemistry, Asymmetric Synthesis, Ligands, Ligands
Other Product Names / Synonyms: (S,S)-ip-FOXAP

Formula: C₂₈H₂₈FeNOP
Melting Pt: 160 °C
MDL Number: MFCD15144869
CAS Number: 163169-29-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4921fc3bc3b420b19f72f5428900aa3899059c3e&vid=4921fc3bc3b420b19f72f5428900aa3899059c3e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR