1,3-Dioxan-2-one ≥98.0%

Supplier: TCI America

D3821-5G D3821-25G
TCD3821-5GEA 366.88 CAD
TCD3821-5G TCD3821-25G
1,3-Dioxan-2-one ≥98.0%
1,3-Dioxan-2-one

CAS Number: 2453-03-4
MDL Number: MFCD00010440
Molecular Formula: C4H6O3
Molecular Weight: 102.09
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 135
Melting point (°C): 48
Flash Point (°C): 110


Product Categories: Materials Chemistry, Monomers, Bioscience, Monomers for Synthetic Polymers (for DDS)
Other Product Names / Synonyms: Trimethylene Carbonate

Formula: C₄H₆O₃
Boiling Pt: 135 °C
Melting Pt: 48 °C
Flash Pt: 110 °C
MDL Number: MFCD00010440
CAS Number: 2453-03-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d151f4fd1565dc7c6bc6a8b85d62f32d6368e23&vid=8d151f4fd1565dc7c6bc6a8b85d62f32d6368e23&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR