Di-mu-chlorobis[5-hydroxy-2-[1-(hydroxyimino)ethyl]phenyl]palladium(II) dimer ≥95.0%
Supplier: TCI America
Synonyms:
Di-μ-chlorobis[5-hydroxy-2-[1-(hydroxyimino-κN)ethyl]phenyl-κC]palladium(II) dimer, Najera Catalyst II
CAS Number: 419581-64-9
MDL Number: MFCD08705289
Molecular Formula: C16H16Cl2N2O4Pd2
Molecular Weight: 584.05
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 255
Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Cross-coupling Reaction using Transition Metal Catalysts
Other Product Names / Synonyms: Najera Catalyst II
Formula:
C₁₆H₁₆Cl₂N₂O₄Pd₂ Melting Pt: 255 °C |
MDL Number:
MFCD08705289 CAS Number: 419581-64-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...