Di-mu-chlorobis[5-chloro-2-[(4-chlorophenyl)(hydroxyimino)methyl]phenyl]palladium(II) dimer ≥95.0%

Supplier: TCI America

Synonyms: Najera Catalyst I

D3806-1G D3806-250MG
TCD3806-1GEA 491.38 CAD
TCD3806-1G TCD3806-250MG
Di-mu-chlorobis[5-chloro-2-[(4-chlorophenyl)(hydroxyimino)methyl]phenyl]palladium(II) dimer ≥95.0%
Di-mu-chlorobis[5-chloro-2-[(4-chlorophenyl)(hydroxyimino)methyl]phenyl]palladium(II) dimer

CAS Number: 287410-78-0
MDL Number: MFCD09038424
Molecular Formula: C26H16Cl6N2O2Pd2
Molecular Weight: 813.97
Purity/Analysis Method: >95.0% (T)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Cross-coupling Reaction using Transition Metal Catalysts
Other Product Names / Synonyms: Najera Catalyst I

Formula: C₂₆H₁₆Cl₆N₂O₂Pd₂
MW: 813.97 g/mol
MDL Number: MFCD09038424
CAS Number: 287410-78-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de456ae193a31fcb213195fa077319984b0e1470&vid=de456ae193a31fcb213195fa077319984b0e1470&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR