2,6-Dibromodithieno[3,2-b:2',3'-d]thiophene ≥98.0%

Supplier: TCI America

D3799-1G D3799-200MG
TCD3799-1GEA 611.62 CAD
TCD3799-1G TCD3799-200MG
2,6-Dibromodithieno[3,2-b:2',3'-d]thiophene ≥98.0%
2,6-Dibromodithieno[3,2-b:2',3'-d]thiophene

CAS Number: 67061-69-2
Molecular Formula: C8H2Br2S3
Molecular Weight: 354.09
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 171

Formula: C₈H₂Br₂S₃
Melting Pt: 171 °C
MDL Number: MFCD11215199
CAS Number: 67061-69-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e726814ca7cf9282bd15afe7de906f04c06cbdb6&vid=e726814ca7cf9282bd15afe7de906f04c06cbdb6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR