4-Amino-4'-nitrobiphenyl ≥96.0%

Supplier: TCI America

Synonyms: 4-Amino-4'-nitrodiphenyl

A1251-1G A1251-5G
TCA1251-001GEA 301.78 CAD
TCA1251-001G TCA1251-005G
4-Amino-4'-nitrobiphenyl ≥96.0%
4-Amino-4'-nitrobiphenyl

CAS Number: 1211-40-1
MDL Number: MFCD00093527
Molecular Formula: C12H10N2O2
Molecular Weight: 214.22
Purity/Analysis Method: >96.0% (T)
Form: Crystal
Melting point (°C): 204


Product Categories: Materials Chemistry, Biphenyls (for High-Performance Polymer Research), Organic Nonlinear Optical (NLO) Materials
Other Product Names / Synonyms: 4-Amino-4'-nitrodiphenyl

Formula: C₁₂H₁₀N₂O₂
Melting Pt: 204 °C
MDL Number: MFCD00093527
CAS Number: 1211-40-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f5d3ab05a44ae609df2fe7e3c1ace98d3977b20&vid=9f5d3ab05a44ae609df2fe7e3c1ace98d3977b20&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR