3,3'-Dibromo-2,2'-bithiophene ≥98.0%

Supplier: TCI America

D3798-1G D3798-5G D3798-25G
TCD3798-1GEA 107.44 CAD
TCD3798-1G TCD3798-5G TCD3798-25G
3,3'-Dibromo-2,2'-bithiophene ≥98.0%
3,3'-Dibromo-2,2'-bithiophene

CAS Number: 51751-44-1
MDL Number: MFCD00114806
Molecular Formula: C8H4Br2S2
Molecular Weight: 324.05
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 99
Storage Temperature: 0-10°C

Formula: C8H4Br2S2
MW: 324.0554 g/mol
Melting Pt: 99 °C
MDL Number: MFCD00114806
CAS Number: 51751-44-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0fc88b595473bcb266daa1b8249d33920e8eccac&vid=0fc88b595473bcb266daa1b8249d33920e8eccac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR