trans-2,5-Dichlorocinnamic acid ≥96.0%
Supplier: TCI America
Synonyms:
trans-3-(2,5-Dichlorophenyl)-2-propenoic Acid
D3794-25G
D3794-5G
TCD3794-25GEA
246.32
CAD
TCD3794-25G
TCD3794-5G
trans-2,5-Dichlorocinnamic acid ≥96.0%
trans-2,5-Dichlorocinnamic acid
CAS Number: 20595-47-5
MDL Number: MFCD03002786
Molecular Formula: C9H6Cl2O2
Molecular Weight: 217.05
Purity/Analysis Method: >96.0% (GC,T)
Form: Crystal
Melting point (°C): 192
Product Categories: Bioscience, Phenylpropanoids
Other Product Names / Synonyms: trans-3-(2,5-Dichlorophenyl)-2-propenoic Acid
Formula:
C₉H₆Cl₂O₂ MW: 217.05 g/mol Melting Pt: 192 °C |
MDL Number:
MFCD03002786 CAS Number: 20595-47-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...