2,6-Dichlorodiphenylamine ≥98.0%

Supplier: TCI America

Synonyms: N-(2,6-Dichlorophenyl)aniline, 2,6-Dichloro-N-phenylaniline

D3778-250G D3778-25G
TCD3778-250GEA 330.54 CAD
TCD3778-250G TCD3778-25G
2,6-Dichlorodiphenylamine ≥98.0%
2,6-Dichlorodiphenylamine

CAS Number: 15307-93-4
MDL Number: MFCD00269648
Molecular Formula: C12H9Cl2N
Molecular Weight: 238.11
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 51
Flash Point (°C): 110


Other Product Names / Synonyms: N-(2,6-Dichlorophenyl)aniline

Formula: C₁₂H₉Cl₂N
MW: 238.11 g/mol
Melting Pt: 51 °C
Flash Pt: 110 °C
MDL Number: MFCD00269648
CAS Number: 15307-93-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7cf6743cdc7a613a819afffaf1ed4f816631402d&vid=7cf6743cdc7a613a819afffaf1ed4f816631402d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR