Diethyl-1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
Synonyms:
Hantzsch Ester, 1,4-Dihydro-2,6-dimethyl-3,5-pyridinedicarboxylic Acid Diethyl Ester, Diethyl 1,4-dihydro-3,5-pyridinecarboxylate, Etidin, Diethyl 1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate
CAS Number: 1149-23-1
MDL Number: MFCD00005951
Molecular Formula: C13H19NO4
Molecular Weight: 253.30
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 183
Lambda max.: 374 nm (EtOH)
Product Categories: Synthetic Organic Chemistry, Reduction
Other Product Names / Synonyms: 1,4-Dihydro-2,6-dimethyl-3,5-pyridinedicarboxylic Acid Diethyl Ester, Hantzsch Ester
Formula:
C₁₃H₁₉NO₄ MW: 253.30 g/mol Melting Pt: 183 °C |
MDL Number:
MFCD00005951 CAS Number: 1149-23-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...