1,1-Dimethyl-3-[3-(trifluoromethyl)phenyl]urea ≥97.0%
Supplier: TCI America
Synonyms:
Fluometuron
D3773-25G
D3773-5G
TCD3773-25GEA
265.44
CAD
TCD3773-25G
TCD3773-5G
1,1-Dimethyl-3-[3-(trifluoromethyl)phenyl]urea ≥97.0%
1,1-Dimethyl-3-[3-(trifluoromethyl)phenyl]urea
CAS Number: 2164-17-2
MDL Number: MFCD00018012
Molecular Formula: C10H11F3N2O
Molecular Weight: 232.21
Purity/Analysis Method: >97.0% (N)
Form: Crystal
Melting point (°C): 162
Product Categories: Synthetic Organic Chemistry, Fluorinated Bioactive Compounds, Bioscience, Herbicide Ingredients
Other Product Names / Synonyms: Fluometuron
Formula:
C₁₀H₁₁F₃N₂O Melting Pt: 162 °C |
MDL Number:
MFCD00018012 CAS Number: 2164-17-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...