3,5-Dimethylisoxazole-4-boronic acid pinacol ester ≥98.0%

Supplier: TCI America

D3772-1G D3772-5G
TCD3772-1GEA 102.7 CAD
TCD3772-1G TCD3772-5G
3,5-Dimethylisoxazole-4-boronic acid pinacol ester ≥98.0%
3,5-Dimethylisoxazole-4-boronic acid pinacol ester

CAS Number: 832114-00-8
MDL Number: MFCD05863910
Molecular Formula: C11H18BNO3
Molecular Weight: 223.08
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 98


Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives
Other Product Names / Synonyms: (3,5-Dimethyl-4-isoxazolyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3,5-Dimethyl-4-isoxazoleboronic Acid Pinacol Ester

Formula: C₁₁H₁₈BNO₃
Melting Pt: 98 °C
MDL Number: MFCD05863910
CAS Number: 832114-00-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c8ab120965ff0a6f8f636b3ab05e70f6a92efce4&vid=c8ab120965ff0a6f8f636b3ab05e70f6a92efce4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR