4-(4-Aminophenyl)benzonitrile ≥95.0%

Supplier: TCI America

Synonyms: 4-Amino-[1,1-biphenyl]-4-carbonitrile, 4-Amino-4'-cyanodiphenyl, 4'-Aminobiphenyl-4-carbonitrile, 4-(4-Aminophenyl)benzonitrile, 4-Amino-4'-cyanobiphenyl

A1249-1G A1249-5G
TCA1249-001GEA 203.82 CAD
TCA1249-001G TCA1249-005G
4-(4-Aminophenyl)benzonitrile ≥95.0%
4-(4-Aminophenyl)benzonitrile

CAS Number: 4854-84-6
MDL Number: MFCD00191367
Molecular Formula: C13H10N2
Molecular Weight: 194.24
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 185
Flash Point (°C): 100


Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: 4'-Aminobiphenyl-4-carbonitrile, 4-Amino-4'-cyanodiphenyl

Formula: C₁₃H₁₀N₂
MW: 194.2319 g/mol
Melting Pt: 185 °C
Flash Pt: 100 °C
MDL Number: MFCD00191367
CAS Number: 4854-84-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f164f2617c57134b86971054d9939f01316a1a3&vid=5f164f2617c57134b86971054d9939f01316a1a3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR