Diethyl-2-propyl-1H-imidazole-4,5-dicarboxylate ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D3765-25G D3765-5G
TCD3765-25GEA 260.7 CAD
TCD3765-25G TCD3765-5G
Diethyl-2-propyl-1H-imidazole-4,5-dicarboxylate ≥98.0% (by GC, titration analysis)
Diethyl-2-propyl-1H-imidazole-4,5-dicarboxylate

CAS Number: 144689-94-1
MDL Number: MFCD08062365
Molecular Formula: C12H18N2O4
Molecular Weight: 254.29
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 86


Other Product Names / Synonyms: 2-Propyl-1H-imidazole-4,5-dicarboxylic Acid Diethyl Ester

Formula: C₁₂H₁₈N₂O₄
Melting Pt: 86 °C
MDL Number: MFCD08062365
CAS Number: 144689-94-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ae8bc16a22c56acf0c33caaafa11e69e2cec9bf&vid=2ae8bc16a22c56acf0c33caaafa11e69e2cec9bf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR