2,3:4,5-Di-O-isopropylidene-β-D-fructopyranose ≥98.0%

Supplier: TCI America

D3758-5G D3758-25G
TCD3758-5GEA 46.63 CAD
TCD3758-5G TCD3758-25G
2,3:4,5-Di-O-isopropylidene-β-D-fructopyranose ≥98.0%
2,3:4,5-Di-O-isopropylidene-beta-D-fructopyranose

CAS Number: 20880-92-6
MDL Number: MFCD00022183
Molecular Formula: C12H20O6
Molecular Weight: 260.29
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 96
Specific rotation [a]20/D: -34 deg (C=1, H2O)
Storage Temperature: <0°C

Formula: C₁₂H₂₀O₆
Melting Pt: 96 °C
MDL Number: MFCD00022183
CAS Number: 20880-92-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b959b280753d1a5a3766b66f4cb998156579d70&vid=3b959b280753d1a5a3766b66f4cb998156579d70&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR