3,3'-Diaminobenzidine tetrahydrochloride hydrate ≥98.0% (by HPLC, total nitrogen) for biochemical research

Supplier: TCI America

D3757-1G D3757-5G
TCD3757-1GEA 76.05 CAD
TCD3757-1G TCD3757-5G
3,3'-Diaminobenzidine tetrahydrochloride hydrate ≥98.0% (by HPLC, total nitrogen) for biochemical research
3,3'-Diaminobenzidine tetrahydrochloride hydrate

[for Biochemical Research]
CAS Number: 868272-85-9
MDL Number: MFCD08273058
Molecular Formula: C12H14N4
Molecular Weight: 360.10
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 280
Flash Point (°C): 200
Storage Temperature: 0-10°C


Product Categories: Bioscience, Stains & Dyes, Peroxidase Substrates
Other Product Names / Synonyms: 3,3',4,4'-Tetraaminobiphenyl Tetrahydrochloride, DAB.4HCl

Formula: C₁₂H₁₄N₄·4HCl·H₂O
Melting Pt: 280 °C
Flash Pt: 200 °C
MDL Number: MFCD08273058
CAS Number: 868272-85-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e7d4920e6188c34250693d918e38eb4206b0213&vid=5ba8d8f0970ab1a54a4251afa4482eae588adcfe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR