3,3'-Diaminobenzidine (DAB) ≥97.0% (by titrimetric analysis) for biochemical research

Supplier: TCI America

D3756-1G D3756-5G
TCD3756-1GEA 52.14 CAD
TCD3756-1G TCD3756-5G
3,3'-Diaminobenzidine (DAB) ≥97.0% (by titrimetric analysis) for biochemical research
3,3'-Diaminobenzidine

[for Biochemical Research]
CAS Number: 91-95-2
MDL Number: MFCD00007725
Molecular Formula: C12H14N4
Molecular Weight: 214.27
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 177
Flash Point (°C): 200
Storage Temperature: 0-10°C


Product Categories: Bioscience, Stains & Dyes, Peroxidase Substrates
Other Product Names / Synonyms: 3,3',4,4'-Tetraaminobiphenyl, DAB

Formula: C₁₂H₁₄N₄
Melting Pt: 177 °C
Flash Pt: 200 °C
MDL Number: MFCD00007725
CAS Number: 91-95-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ccfb33f1daa3428fc9becddaa33e9f70fcdd5fe6&vid=efac03bb94ef3767610c7242f336f8d22432f0dc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR