2-Acetamido-6-hydroxypurine ≥95.0% (by HPLC, titration analysis)
Supplier: TCI America
A1248-1G
A1248-5G
TCA1248-1GEA
340.49
CAD
TCA1248-1G
TCA1248-5G
2-Acetamido-6-hydroxypurine ≥95.0% (by HPLC, titration analysis)
2-Acetamido-6-hydroxypurine
CAS Number: 19962-37-9
MDL Number: MFCD00078201
Molecular Formula: C7H7N5O2
Molecular Weight: 193.17
Purity/Analysis Method: >95.0% (HPLC,T)
Form: Crystal
Storage Temperature: <0°C
Product Categories: Bioscience, Nucleobases and Their Analogs
Other Product Names / Synonyms: N-Acetylguanine
Formula:
C₇H₇N₅O₂ |
MDL Number:
MFCD00078201 CAS Number: 19962-37-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...