1,2-Di(4-pyridyl)ethane ≥98.0%

Supplier: TCI America

Synonyms: 4,4'-Ethylenedipyridine, 1,2-Di(pyridin-4-yl)ethane, 1,2-Bis(4-pyridyl)ethane

D3752-1G D3752-5G
TCD3752-1GEA 71.1 CAD
TCD3752-1G TCD3752-5G
1,2-Di(4-pyridyl)ethane ≥98.0%
1,2-Di(4-pyridyl)ethane

CAS Number: 4916-57-8
MDL Number: MFCD00006451
Molecular Formula: C12H12N2
Molecular Weight: 184.24
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 174
Melting point (°C): 112


Product Categories: Materials Chemistry, Metal Organic Framework (MOF) and Porous Coordination Polymer (PCP) Research Reagents
Other Product Names / Synonyms: 1,2-Bis(4-pyridyl)ethane, 4,4'-Ethylenedipyridine

Formula: C₁₂H₁₂N₂
MW: 184.24 g/mol
Boiling Pt: 174 °C
Melting Pt: 112 °C
MDL Number: MFCD00006451
CAS Number: 4916-57-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae42820e25fcab2668b6cc5aba677fdfb343a409&vid=ae42820e25fcab2668b6cc5aba677fdfb343a409&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR