3,6-Dichlorocarbazole ≥96.0%

Supplier: TCI America

D3751-1G D3751-5G
TCD3751-1GEA 82.63 CAD
TCD3751-1G TCD3751-5G
3,6-Dichlorocarbazole ≥96.0%
3,6-Dichlorocarbazole

CAS Number: 5599-71-3
MDL Number: MFCD00218271
Molecular Formula: C12H7Cl2N
Molecular Weight: 236.10
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Melting point (°C): 208

Formula: C₁₂H₇Cl₂N
Melting Pt: 208 °C
MDL Number: MFCD00218271
CAS Number: 5599-71-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2f3a97e323b51b10efe941bf5b64d5be12f2571&vid=e2f3a97e323b51b10efe941bf5b64d5be12f2571&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR