Dimethyl decahydro-2,6-naphthalenedicarboxylate (mixture of isomers) ≥98.0%

Supplier: TCI America

D3745-25G D3745-5G
TCD3745-25GEA 1262.42 CAD
TCD3745-25G TCD3745-5G
Dimethyl decahydro-2,6-naphthalenedicarboxylate (mixture of isomers) ≥98.0%
Dimethyl decahydro-2,6-naphthalenedicarboxylate (mixture of isomers)

CAS Number: 3068-02-8
Molecular Formula: C14H22O4
Molecular Weight: 254.33
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 140


Other Product Names / Synonyms: Decahydro-2,6-naphthalenedicarboxylic Acid Dimethyl Ester

Formula: C₁₄H₂₂O₄
Boiling Pt: 140 °C
CAS Number: 3068-02-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=efd8371092aae29e1abedeab6cf910adaa65658d&vid=efd8371092aae29e1abedeab6cf910adaa65658d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR